Drain Hose for 1L or 4.5L FreeZone Freeze Dry System
Actual product may vary
Drain Hose for 1L or 4.5L FreeZone Freeze Dry System
Labconco™ Drain Hose for 1L or 4.5L FreeZone Freeze Dry System is a replacement hose that allows moisture from condensation to be emptied from the freeze dryer.
Have Questions?
Catalog Number
For Use With
LBC7645904
1L or 4.5L FreeZone System
Catalog number LBC7645904
Price (MYR)
-
For Use With:
1L or 4.5L FreeZone System
FreeZone Freeze Dryers
Designed to handle the lyophilization needs of research and pilot plant laboratories
All sizes/models include a large touch screen display
Lyo-Works™ OS
Provides a real time display of temperature and vacuum level
Optional End-Zone™ end point detection system
Console models are offered in a range of sizes with optional stoppering tray
Collectors that reach -50°C are suitable for lyophilizing aqueous samples
Collectors that reach -84°C are ideal for freeze drying samples with low eutectic temperatures, including ones containing acetonitrile
Specifications
MaterialRubber
For Use With1L or 4.5L FreeZone System
For Use With (Application)Removing condensation moisture