Specifications
ColorSilver
For Use With (Application)Fits 5mL, 10mL, 20mL, 30mL, 50mL, 100mL and 125mL Serum bottles
MaterialAluminum
SterilityNon-sterile
Weight (Metric)0.5kg
Weight (English)1 lb.
Thread Size (Metric)57 mm
Neck Finish57 mm
AutoclavableAutoclavable
DisposableSingle-use
Product TypeCap
Unit SizePK