Labconco 2 ml Pre-Scored Flat Bottom Ampule
Labconco 2 ml Pre-Scored Flat Bottom Ampule
Actual product may vary
Labconco 2 ml Pre-Scored Flat Bottom Ampule
Labconco Ampules are fabricated of highest quality borosilicate glass for strength and durability.
Have Questions?
Catalog NumberCapacity (Metric)
LBC75708102 mL
Catalog number LBC7570810
Price (MYR)
2,733.00
EA
Add to cart
Capacity (Metric):
2 mL
Price (MYR)
2,733.00
EA
Add to cart
Labconco Ampules are fabricated of highest quality borosilicate glass for strength and durability. Ampules are sold in packages of 100.
Specifications
Capacity (Metric)2 mL
MaterialBorosilicate Glass
Weight (English)3.0 lbs
Weight (Metric)1.4 kg
Unit SizeEA