Mini Stoppering Chamber
Mini Stoppering Chamber
Actual product may vary
Mini Stoppering Chamber
For lyophilizing samples in small serum vials with split stoppers
Have Questions?
Catalog number LBC7569000
Price (MYR)
12,513.00
EA
Add to cart
Price (MYR)
12,513.00
EA
Add to cart
  • For use with a FreeZone™ Freeze Dry System
  • Accommodates 48 serum vials of 2 mL and 8 serum vials of 30 mL
  • After lyophilization, vials can be stoppered while still under vacuum
  • Stoppering travel distance: 0.5 in. (1.3 cm)
  • For vials ranging in height from 1.38 to 2.48 in. (3.5 to 6.3 cm)
  • Comes with tray for loading and unloading of samples
Specifications
TypeMini Stoppering Chamber
No. of Shelves0
MaterialAcrylic
For Use With (Application)For lyophilizing samples in small serum vials with split stoppers
For Use With (Equipment)FreeZone™ Freeze Dry System, 48 serum vials (2 mL) or 8 serum vials (30 mL)
Includes17 in. of clear pvc tubing
Width (English)12 in.
Width (Metric)30.4 cm
Height (English)10 in.
Height (Metric)25.4 cm
Weight (English)15 lb.
Weight (Metric)6.8 kg
Unit SizeEA