Check Order Status
Fast-Freeze™ Flasks, Rubber Tops Only
Actual product may vary
Fast-Freeze™ Flasks, Rubber Tops Only
For use with Fast-Freeze Flasks
Have Questions?
Catalog Number
Capacity (Metric)
Diameter (Metric) Flask Top Adapter
LBC7544400
900 mL
1.9 cm
Catalog number
LBC7544400
Price (MYR)
920.00
EA
Add to cart
Capacity (Metric):
900 mL
Diameter (Metric) Flask Top Adapter:
1.9 cm
Price (MYR)
920.00
EA
Add to cart
For use with Fast-Freeze Flasks
Flexible, noncontaminating silicone rubber top Snaps on and off easily, yet provides a reliable, high-vacuum seal
Specifications
Diameter (Metric) Flask Top Adapter
1.9 cm
Capacity (Metric)
900 mL
Material
Borosilicate Glass
Product Type
Flask Top
Weight (English)
1 lb.
Disposable
Reusable
Mouth Style
Wide
Bottom Shape
Flat
Diameter (Metric) Outer
11.1 cm
Weight (Metric)
0.5 kg