Aseptic Valved Flask Adapter
Aseptic Valved Flask Adapter
Actual product may vary
Aseptic Valved Flask Adapter
For use with Fast-Freeze Flasks
Have Questions?
Catalog number LBC7543900
Price (MYR)
1,317.00
EA
Add to cart
Price (MYR)
1,317.00
EA
Add to cart
Aseptic valved flask adapter allows filling user-supplied Labconco Fast-Freeze Flask under sterile conditions prior to lyophilization.
  • For use with Labconco Fast-Freeze Flasks 120 mL up to 2000 mL.
  • Valve closes to isolate contents for transportation.
Specifications
TypeAseptic Valved Flask Adapter
ColorWhite
Width (English)1.75 in.
Width (Metric)4.4 cm
Height (English)1.75 in.
Height (Metric)4.4 cm
Length (English)3.15 in.
Length (Metric)8 cm
Shipping Weight (Metric)226 g
Unit SizeEA