Fast-Freeze™ Flasks, Flask Bottom Only
Fast-Freeze™ Flasks, Flask Bottom Only
Actual product may vary
Fast-Freeze™ Flasks, Flask Bottom Only
High-strength borosilicate glass bottom to withstand extreme temperatures and high vacuum.
Have Questions?
Catalog NumberCapacity (Metric)
LBC7542800600 mL
Catalog number LBC7542800
Price (MYR)
1,329.00
EA
Add to cart
Capacity (Metric):
600 mL
Price (MYR)
1,329.00
EA
Add to cart
High-strength borosilicate glass bottom to withstand extreme temperatures and high vacuum
  • Features a wide mouth opening to load fast and clean easily
  • Flat bottom reduces spills and provides better balance
  • Flasks should be filled to no more than one-third of their volume so that maximum surface area is achieved and efficient lyophilization is assured
  • Tops, bottoms, and filter paper are available separately as replacement components
Specifications
Diameter (English) Outer3.55 in.
Capacity (Metric)600 mL
MaterialBorosilicate Glass
Height (English)5.3 in.
Height (Metric)13.5 cm
Product TypeFlask
Weight (English)1.8 lb.
DisposableReusable
Mouth StyleWide
Bottom ShapeFlat
Diameter (Metric) Outer90.2 mm
Weight (Metric)0.8 kg
Unit SizeEA