Fast-Freeze™ Flasks, Complete Assembly
Fast-Freeze™ Flasks, Complete Assembly
Actual product may vary
Fast-Freeze™ Flasks, Complete Assembly
Fast-Freeze™ Flasks are easy to handle and fast to load.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
LBC75412002000 mL
LBC754000040 mL
LBC7540400150 mL
LBC7540600300 mL
LBC7540800600 mL
LBC75410001200 mL
LBC7540900900 mL
LBC7541100750 mL
Catalog number LBC7541200
Price (MYR)
3,540.00
EA
Add to cart
Capacity (Metric):
2000 mL
Price (MYR)
3,540.00
EA
Add to cart
Easy to handle and fast to load
  • Flat borosilicate glass
  • Silicone rubber snap-on tops provide reliable high-vacuum seals
  • Eliminate risk of contamination by vacuum grease; reduce spillage and sample loss
  • No easily misplaced small parts
  • Wide openings
  • Should be filled no more than one-third of the volume so a maximum surface area is achieved
  • Compatible with all major brands of freeze-drying equipment, including Labconco and Virtis™
Specifications
Diameter (English) Outer3.55 in.
Diameter (Metric) Flask Top Adapter1.9 cm
Capacity (Metric)2000 mL
MaterialBorosilicate Glass
For Use With (Equipment)Compatible with all major brands of freeze-drying equipment
Height (English)14.9 in.
Height (Metric)38 cm
Product TypeFlask
Weight (English)2.2 lb.
DisposableReusable
Mouth StyleWide
Bottom ShapeFlat
Diameter (Metric) Outer90.2 mm
Weight (Metric)1 kg
Unit SizeEA