4-Port Manifold for FreeZone™ Freeze Dryers
4-Port Manifold for FreeZone™ Freeze Dryers
Actual product may vary
4-Port Manifold for FreeZone™ Freeze Dryers
Stainless steel manifold for use with FreeZone™ Freeze Dryers includes neoprene valves with plastic knobs.
Have Questions?
Catalog number LBC7201700
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Stainless steel manifold for use with FreeZone™ Freeze Dryers includes neoprene valves with plastic knobs.
4-Port Manifold
  • Type 304 stainless steel manifold
  • 4 neoprene valves with plastic knobs accommodate both 1/2 in. and 3/4 in. adapters
  • For use with 1 L, 2.5L, 4.5 L, 6L, 8L, 12 L, or 18 L FreeZone Freeze Dryers
  • Freeze Dryer, glassware, and adapters are required (not included)
  • Includes one latching pin
Specifications
TypeDrying Manifold
No. of Shelves0
MaterialStainless Steel (304)
No. of Ports4
For Use With (Equipment)1 L, 2.5 L, 4.5 L, 6 L, 8 L, 12 L, or 18 L FreeZone™ Freeze Dryers
Includes4 neoprene valves with molded plastic knobs and 1 latching pin
Width (English)8.6 in.
Height (English)11.2 in.
Length (English)8.6 in.
Shipping Weight (English)5.5 lb.
Weight (English)4 lb.
Unit SizeEA