FreeZone™ 4.5 L -84°C Benchtop Freeze Dryer, 230 V Model
FreeZone™ 4.5 L -84°C Benchtop Freeze Dryer, 230 V Model
Actual product may vary
FreeZone™ 4.5 L -84°C Benchtop Freeze Dryer, 230 V Model
FreeZone 4.5 Liter Benchtop Freeze Dryers are designed for lyophilizing light to moderate sample loads. The compact design is ideal for labs with limited floor space. New Lyo-Works™ OS provides flexibility and convenience in an easy-to-use, touch screen display.
Have Questions?
Catalog NumberIncludesPlug Type
LBC710401050Non coated Stainless steel collectorBritish (UK)
Catalog number LBC710401050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Includes:
Non coated Stainless steel collector
Plug Type:
British (UK)
FreeZone 4.5 Liter Benchtop Freeze Dryers are designed for lyophilizing light to moderate sample loads. The compact design is ideal for labs with limited floor space. New Lyo-Works™ OS provides flexibility and convenience in an easy-to-use, touch screen display.
  • Upright stainless steel collector coil holds 4.5 liters of ice before defrosting
  • --84° C collector is suitable for low eutectic point samples including acetonitrile
  • Features include hot gas defrost, side-mounted USB port and rear-mounted Ethernet connection
  • Environmentally-friendly HCFC/CFC-free refrigeration system
  • Internal coil maximizes vapor-trapping efficiency
  • Optional PTFE-coated collectors withstand corrosive compounds
  • Lyo-Works OS provides a real time display of collector temperature and vacuum level
  • 5 in. full color touch screen display
  • Flexible programming and control of optional drying accessories
  • Optional End-Zone™ end point detection for flask freeze drying
  • One-touch data logging and display provides information to maximize sample quality and minimize run time
  • Customizable settings include language, time zone, temperature and vacuum
  • Alerts and feedback of current operating parameters can be sent automatically to user-entered email addresses
  • FreeZone exclusive features protect your vacuum pump from costly damage: A vacuum break valve bleeds air into the system when power to the freeze dryer or vacuum pump is shut off for more than 5 minutes. A moisture sensor prevents refrigeration or vacuum start-up when moisture is detected in the drain line. Vacuum pump is automatically shut down when a leak is present in initial pull down or during run
Vacuum pump with a displacement of at least 98 liters/minute, drying accessory and freeze dry glassware (if not bulk freeze drying) are required (not included).
Specifications
TypeFreeze Dryer/Lyophilizer
Capacity (Metric)2.5 L (before defrosting)
No. of Shelves0
Temperature Range (Metric)-84°C
Temperature Range (English)-119°C
MaterialSteel
Certifications/ComplianceUL, CAN/CSA, CE
IncludesNon coated Stainless steel collector
Depth (English)26.5 in.
Depth (Metric)67.3 cm
Width (English)22.5 in.
Width (Metric)57.1 cm
Height (English)18.5 in.
Height (Metric)46.9 cm
Amperage10 A
Plug TypeBritish (UK)
Voltage230 V
Frequency60 Hz
Shipping Weight (English)166 lb.
Shipping Weight (Metric)75.3 kg
Unit SizeEA