FreeZone™ 18L -50°C Console Freeze Dryers, Purge Valve and Mini Chamber
FreeZone™ 18L -50°C Console Freeze Dryers, Purge Valve and Mini Chamber
Actual product may vary
FreeZone™ 18L -50°C Console Freeze Dryers, Purge Valve and Mini Chamber
FreeZone™ 18L Console Freeze Dryers are designed for lyophilizing moderate to large sample loads or numerous small container batches. New Lyo-Works™ OS provides flexibility and convenience in an easy-to-use, touch screen display. Your samples are protected with remote notification and data recording.
Have Questions?
Catalog NumberPlug TypeIncludes
LBC701812250British (UK)PTFE coated collector, Purge Valve, Mini Vacuum Drying Chamber
Catalog number LBC701812250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Plug Type:
British (UK)
Includes:
PTFE coated collector, Purge Valve, Mini Vacuum Drying Chamber
FreeZone™ 18L Console Freeze Dryers are designed for lyophilizing moderate to large sample loads or numerous small container batches. New Lyo-Works™ OS provides flexibility and convenience in an easy-to-use, touch screen display. Your samples are protected with remote notification and data recording.
  • Upright stainless steel collector coil holds 18L of ice before defrosting
  • -50°C collector is suitable for aqueous samples
  • Environmentally-friendly HCFC/CFC-free refrigeration system
  • Internal coil maximizes vapor-trapping efficiency
  • Optional PTFE-coated collectors withstand corrosive compounds
  • Features include hot gas defrost, side-mounted USB port and rear-mounted Ethernet connection. The console provides interior space for vacuum pump and accommodates the FreeZone™ Stoppering Tray Dryer (pump and tray dryer sold separately)
  • Lyo-Works OS provides real time display of collector temperature and vacuum level
  • 5 in. full color touch screen display
  • Flexible programming and control of optional drying accessories
  • Optional End-Zone™ end point detection for flask freeze drying
  • One-touch data logging and display provides information to maximize sample quality and minimize run time
  • Customizable settings include language, time zone, temperature and vacuum
  • Alerts and feedback of current operating parameters can be sent automatically to user-entered email addresses
  • Front viewing window makes monitoring vacuum pump oil hassle free
  • FreeZone™ exclusive features protect your vacuum pump from costly damage: A vacuum break valve bleeds air into the system when power to the freeze dryer or vacuum pump is shut off for more than 5 minutes. A moisture sensor prevents refrigeration or vacuum start-up when moisture is detected in the drain line. Vacuum pump is automatically shut down when a leak is present in initial pull down or during run.
  • Includes a purge valve for isolating the vacuum pump from the freeze dryer and a built-in stainless steel mini vacuum drying chamber with control to +60°C
Vacuum pump with a displacement of at least 98L/min, drying accessory and freeze dry glassware (if not bulk freeze drying) are required (not included).
Specifications
TypeFreeze Dryer/Lyophilizer
Capacity (Metric)18 L (before defrosting)
No. of Shelves0
Temperature Range (Metric)-50°C
Temperature Range (English)-58°F
MaterialSteel
Certifications/ComplianceUL, CAN/CSA, CE
IncludesPTFE coated collector, Purge Valve, Mini Vacuum Drying Chamber
Depth (English)29.6 in.
Depth (Metric)75.1 cm
Width (English)31.8 in.
Width (Metric)80.7 cm
Height (English)36.6 in.
Height (Metric)92.9 cm
Plug TypeBritish (UK)
Voltage230 V
Frequency60 Hz
Shipping Weight (English)305 lb.
Shipping Weight (Metric)138.34 kg
Unit SizeEA