FreeZone™ 6L -50°C Console Freeze Dryers, 230V Models
FreeZone™ 6L -50°C Console Freeze Dryers, 230V Models
Actual product may vary
FreeZone™ 6L -50°C Console Freeze Dryers, 230V Models
FreeZone 6L Console Freeze Dryers are designed for lyophilizing moderate to large sample loads or numerous small container batches. New Lyo-Works™ OS provides flexibility and convenience in an easy-to-use, touch screen display.
Have Questions?
Catalog NumberIncludesPlug Type
LBC700611050Non coated Stainless steel collectorBritish (UK)
Catalog number LBC700611050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Includes:
Non coated Stainless steel collector
Plug Type:
British (UK)
FreeZone 6L Console Freeze Dryers are designed for lyophilizing moderate to large sample loads or numerous small container batches. New Lyo-Works™ OS provides flexibility and convenience in an easy-to-use, touch screen display.
Features include hot gas defrost, side-mounted USB port and rear-mounted Ethernet connection. The console provides interior space for a vacuum pump and accommodates the FreeZone Stoppering Tray Dryer (pump and tray dryer sold separately).
These models offer the broadest range of built-in options including a shell freezer.
  • Upright stainless steel collector coil holds 6 liters of ice before defrosting
  • -50° C collector is suitable for aqueous samples
  • Environmentally-friendly HCFC/CFC-free refrigeration system
  • Internal coil maximizes vapor-trapping efficiency
  • Optional PTFE-coated collectors withstand corrosive compounds
  • Lyo-Works OS provides a real time display of collector temperature and vacuum level
  • 5 in. full color touch screen display
  • Flexible programming and control of optional drying accessories
  • Optional End-Zone™ end point detection for flask freeze drying
  • One-touch data logging and display provides information to maximize sample quality and minimize run time
  • Customizable settings include language, time zone, temperature and vacuum
  • Alerts and feedback of current operating parameters can be sent automatically to user-entered email addresses
  • Front viewing window makes monitoring vacuum pump oil hassle free
  • FreeZone exclusive features protect your vacuum pump from costly damage: A vacuum break valve bleeds air into the system when power to the freeze dryer or vacuum pump is shut off for more than 5 minutes. A moisture sensor prevents refrigeration or vacuum start-up when moisture is detected in the drain line. Vacuum pump is automatically shut down when a leak is present in initial pull down or during run.
  • Options include: purge valve for isolating the vacuum pump from the freeze dryer, built-in stainless steel mini vacuum drying chamber with control to +60° C, and built-in shell freezer with stainless steel bath that includes HCFC/CFC-free refrigeration system capable of cooling the heat transfer solution to -40° C
Vacuum pump with a displacement of at least 98 liters/minute, drying accessory and freeze dry glassware (if not bulk freeze drying) are required (not included).
Specifications
TypeFreeze Dryer/Lyophilizer
Capacity (Metric)8 L (before defrosting)
No. of Shelves0
Temperature Range (Metric)-50°C
Temperature Range (English)-58°F
MaterialSteel
Certifications/ComplianceUL, CAN/CSA, CE
IncludesNon coated Stainless steel collector
Depth (English)29.6 in.
Depth (Metric)75.1 cm
Width (English)31.8 in.
Width (Metric)80.7 cm
Height (English)36.6 in.
Height (Metric)92.9 cm
Amperage8A
Plug TypeBritish (UK)
Voltage230 V
Frequency60 Hz
Shipping Weight (English)255 lb.
Shipping Weight (Metric)115.7 kg
Unit SizeEA