Kjeldahl Micro Digestor
Kjeldahl Micro Digestor
Actual product may vary
Kjeldahl Micro Digestor
Individual electric heaters with adjustable controls
Have Questions?
Catalog NumberAmperageVoltage
LBC60300016 A230 V
Catalog number LBC6030001
Price (MYR)
-
Amperage:
6 A
Voltage:
230 V
Individual electric heaters with adjustable controls
Each 200w heater consists of tubular steel heater element and stainless-steel cup recessed into top insulating board.
  • Accommodates 100mL flasks
  • Six-port glass manifold exhausts fumes
  • Serrated connector for water aspirator
  • Well ventilated to prevent heat accumulation
  • Top is 1.50 in. thick (3.8cm) insulating board; protects wiring and controls
  • Heavy-gauge epoxy-coated aluminum case
Specifications
Product TypeMicro Digestor
DisplayNo Display
Amperage6 A
Hertz50/60 Hz
Voltage230 V
Wattage200 W
Depth (English)9 in.
Depth (Metric)23 cm
Diameter (Metric) Inner57.3 cm
Diameter (Metric) Outer57.3 cm
Dimensions (L x W x H)57.33 x 22.86 x 35.56
Electrical Requirements230 V, 50/60 Hz
For Use With (Equipment)Kjeldahl digestions
Height (English)14 in.
Height (Metric)36 cm
IncludesManifold, two rear brackets and six 100ml flasks
Quantity1 Ea.
Width (English)22.5 in.
Unit SizeEach