FlaskScrubber™ Glassware Washer
FlaskScrubber™ Glassware Washer
Actual product may vary
Labconco
FlaskScrubber™ Glassware Washer
Provide thorough, efficient, and economical cleaning that meets demanding laboratory standards by outfitting your lab with a Labconco FlaskScrubber™ Glassware Washer.
Have Questions?
Catalog NumberElectrical RequirementsIncludesInlet Water Temperature (Metric) Max.Product Type
LBC4420421230 V 50/60 HzOne spindle rack93°CGlassware Washer
Catalog number LBC4420421
Price (MYR)
0.00
EA
Add to cart
Electrical Requirements:
230 V 50/60 Hz
Includes:
One spindle rack
Inlet Water Temperature (Metric) Max.:
93°C
Product Type:
Glassware Washer
Price (MYR)
0.00
EA
Add to cart
Provide thorough, efficient, and economical cleaning that meets demanding laboratory standards by outfitting your lab with a Labconco FlaskScrubber™ Glassware Washer.
Ideal for college and university, government, pharmaceutical, and industrial labs or any facility utilizing reusable glassware. Choose from single-rack FlaskScrubber series models or dual-rack SteamScrubber models. Accessory spindle and open racks are designed specifically for laboratory glassware and are interchangeable between models.
Operating Features
  • Achieve higher water temperatures for enhanced washing, faster drying, and sanitizing
  • LCD information center for customizing cycles and monitoring operations, including internal water temperature (°C or °F) and alarms
  • High water circulation rates for thorough cleaning: 96 gal./min. (363L/min.) when operated on 115V 60Hz, and 112 gal./min. (424L/min.) on 230V 50/60Hz
  • Dual pumps: one for washing and one for draining, to reduce potential for cross-contamination
  • Built-in purified water pump brings unpressurized or pressurized purified water into tank for up to six pure water rinses
  • Built-in steam generator produces hot vapor to penetrate and remove dried residue for cleaner glassware
  • Manual-fill dispensers for detergent (powder or liquid) or rinse aid solution
  • Two user-set cycle programs
  • User-set delayed start (up to 8 hours)
  • Optional tempered safety glass viewing window with 25w interior light
  • Environmentally Friendly: Item contains environmental attributes or features (e.g. water conserving, lead-free, etc.) as described in the item detail
Structural Features
  • Type 304 brushed stainless-steel door, tank, and upper and lower wash arms
  • Freestanding models have stainless-steel exterior sides and top
  • One-piece, stainless-steel, fine mesh particle drain filter traps debris in tank to protect pumps
  • Four leveling feet
  • 0.375 in. female NPT inlet fitting for hot water tap connection
  • Factory-installed drain hose and band clamp
  • Outfitted for washing narrow-necked flasks
  • Supplied with one Type 304 stainless-steel spindle rack with 36 detachable large spindles, 18 holders, 36 clips, and 10 interchangeable small spindles
  • Built-in forced-air drying through the spindles and throughout the tank up to 99 min. and programmable from 38° to 70°C (100° to 150°F)
  • Factory-set cycle programs: For 208/230V models, 10 (add INTENSE, INTENSE PLUS and EXTREME)
Specifications
DisplayDigital
MaterialStainless Steel (304)
Product TypeGlassware Washer
Flow Rate4.7 L/min. for hot tap water, 3.4 L/min. for purified water
Max. Pressure20 to 120 psi for hot tap water pressure at washer
Min. Purified Water Pressure0 psi
Water Consumption13L/Fill
Product LineFlaskScrubber
StyleFreestanding
Certifications/ComplianceUL 61010-1, CAN CSA C22.2 No. 61010.1, ETL, CE
For Use With (Application)Labware/reusable glassware washing
IncludesOne spindle rack
Inlet Water Temperature (English) Min.120°F
Inlet Water Temperature (English) Recommended140°F
Inlet Water Temperature (English) Max.199°F
Inlet Water Temperature (Metric) Max.93°C
Inlet Water Temperature (Metric) Min.49°C
Inlet Water Temperature (Metric) Recommended60°C
Depth (English)27.5 in.
Depth (Metric)69.9 cm
Height (English)34.2 to 36.2 in.
Height (Metric)86.9 to 91.9 cm
Width (English)24.2 in.
Width (Metric)61.5 cm
Weight (English) Shipping239 lb.
Weight (Metric) Shipping108 kg
Amperage12 A
Electrical RatingDomestic/international
Electrical Requirements230 V 50/60 Hz