FlaskScrubber™ Glassware Washers
Actual product may vary
FlaskScrubber™ Glassware Washers
FlaskScrubber™ Glassware Washers
FlaskScrubber™ Glassware Washers
FlaskScrubber™ Glassware Washers
Labconco
FlaskScrubber™ Glassware Washers
Versatile and efficient glassware washers designed for efficient and consistent washing of narrow-neck glassware and general labware.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionElectrical RequirementsStyle
LBC412001013FlaskScrubber Glassware Washer230 V, 50/60 Hz, 16.5 A, 3 PFreestanding Washer
LBC412001010FlaskScrubber Glassware Washer230 V, 50/60 Hz, 11 A, 1 PFreestanding Washer
LBC412101010FlaskScrubber Glassware Washer230 V, 50/60 Hz, 11 A, 1 PFreestanding Washer with Window
Catalog number LBC412001013
Price (MYR)
102,563.00
Each
Description:
FlaskScrubber Glassware Washer
Electrical Requirements:
230 V, 50/60 Hz, 16.5 A, 3 P
Style:
Freestanding Washer
Price (MYR)
102,563.00
Each
Labconco FlaskScrubber™ Glassware Washers are designed for efficient and consistent washing of narrow-neck glassware and general purpose labware. The CleanWorks OS software is accessible through a full-color touch screen interface and provides intuitive and clear monitoring of wash loads – ensuring repeatable, effective and traceable cleaning.
FlaskScrubber models reach a maximum of 93° C wash temperature and quickly dry glassware with standard forced air spindle injection drying with SpeedVent heat-management technology. FlaskScrubber models include a 36-place lower spindle rack, and may be outfitted with a wide variety inserts and additional racks to accommodate all types of labware.
  • Touch screen display with intuitive CleanWorks OS software
  • Flexible design allows for use of Spindle Injection and Standard racks
  • Max wash temperature 93° C removes tough contaminants
  • Advanced wash pump maximizes water circulation up to 112 gal/min (424 liters/min)
  • Standard forced air spindle drying with SpeedVent technology
  • Durable Type 304 stainless steel chamber and racks
  • 36-place Spindle Injection Rack included
Specifications
DescriptionFlaskScrubber Glassware Washer
DisplayFull-color Touch Screen
MaterialStainless Steel (304)
Product TypeLaboratory Glassware Washer
Flow RateHot water inlet 1.25 GPM (4.7 LPM), Cold water inlet of 3.5 GPM (13.2 LPM)
Max. Pressure120 psi (inlet)
Min. Purified Water PressureN/A, can be supplied from pressurized and non-pressurized sources
Volume (English)6.73 ft.3 chamber volume
Volume (Metric)190.6 L chamber volume
Wash Cycles9 programs with 1 to 9 total cycles
Water ConsumptionAs low as 12.9 L
StyleFreestanding Washer
Certifications/ComplianceUL61010, CAN/CSA C22.2, CE
For Use With (Application)Ideal for College and University, Government, Pharmaceutical and Industrial Labs or Any Facility Utilizing Reusable Glassware
IncludesLower spindle rack
Inlet Water Temperature (English) Min.120°F
Inlet Water Temperature (English) Recommended120°F
Inlet Water Temperature (English) Max.160°F
Inlet Water Temperature (Metric) Max.71°C
Inlet Water Temperature (Metric) Min.49°C
Inlet Water Temperature (Metric) Recommended49°C
Depth (English)27.8 in.
Depth (Metric)70.6 cm
Dimensions (D x W x H) Interior27.8 x 24.3 x 32.2 to 36.2 in.
Height (English)32.2 to 36.2 in.
Height (Metric)81.8 to 91.9 cm
Width (English)24.3 in.
Width (Metric)61.7 cm
Weight (English) Shipping230 lb.
Weight (Metric) Shipping104.3 kg
Amperage16.5 A
Electrical Requirements230 V, 50/60 Hz, 16.5 A, 3 P
Voltage208/230 V