Ultraviolet Lamp Kit for Labconco™ Purifier™ Class II Safety Cabinets
Ultraviolet Lamp Kit for Labconco™ Purifier™ Class II Safety Cabinets
Actual product may vary
Ultraviolet Lamp Kit for Labconco™ Purifier™ Class II Safety Cabinets
Have Questions?
Catalog NumberFor Use With (Equipment)
LBC3745001Purifier Delta Biosafety Cabinet (72 in.)
Catalog number LBC3745001
Price (MYR)
2,288.00
EA
Add to cart
For Use With (Equipment):
Purifier Delta Biosafety Cabinet (72 in.)
Price (MYR)
2,288.00
EA
Add to cart
With 254nm UV lamp and installation instructions.
Specifications
For Use With (Equipment)Purifier Delta Biosafety Cabinet (72 in.)
Weight (English)5 lb.
Weight (Metric)2.3 Kg
Includes254 nm UV Lamp and Installation Instructions
Product TypeConversion Kit
Unit SizeEA