Check Order Status
PrimeMate™ Vacuum Pump Oil
Actual product may vary
Labconco
PrimeMate™ Vacuum Pump Oil
For use with the Labconco PrimeMate Oil Change System
Have Questions?
Catalog Number
Material
LBC1988001
Hydro-treated Paraffinic Hydrocarbon Oil
Catalog number
LBC1988001
Price (MYR)
2,288.00
EA
Add to cart
Material:
Hydro-treated Paraffinic Hydrocarbon Oil
Price (MYR)
2,288.00
EA
Add to cart
Choice of one gallon (3.8L) hydro-treated paraffinic hydrocarbon oil or synthetic hydrocarbon oil
Recommended for Labconco general purpose vacuum pumps
Specifications
Product Type
Vacuum Pump Oil
Format
Oil
For Use With (Equipment)
PrimeMate Oil Change System
Material
Hydro-treated Paraffinic Hydrocarbon Oil
Volume (English)
1 gal.
Volume (Metric)
3.8 L