Kimtech Pure™ W4 Wipers
Kimtech Pure™ W4 Wipers
Actual product may vary
Kimtech Pure™ W4 Wipers
For ISO Class 4 and higher applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W)Product TypePackaging QuantitySterility
KLC7649011 x 9 in. (28 x 23 cm)Pre-saturated Wipe40 Sheets/Pk.Sterile
FIS06-666-1212 x 12 in.Dry Wipe100 Sheets/Pk.Non-sterile
KLC3333012 x 12 in.Dry Wipe100 Sheets/Pk.Non-sterile
Catalog number KLC76490
Price (MYR)
-
Dimensions (L x W):
11 x 9 in. (28 x 23 cm)
Product Type:
Pre-saturated Wipe
Packaging Quantity:
40 Sheets/Pk.
Sterility:
Sterile
  • Made of 100% polypropylene that resists acids, bases and solvents
  • Thermally bonded—no glues or binders
  • Straight, clean, knife-cut edges minimize lint
Specifications
Product TypePre-saturated Wipe
Length (English)11 in.
Length (Metric)28 cm
Width (English)9 in.
Width (Metric)23 cm
MaterialPolypropylene
ColorWhite
EdgesStraight clean knife-cut
Dimensions (L x W)11 x 9 in. (28 x 23 cm)
For Use With (Application)For ISO Class 4 and higher cleanroom applications
Packaging Quantity40 Sheets/Pk.
Saturation Solution70% Isopropyl Alcohol, 30% Deionised Water