G3 Sterile White Nitrile Hand Specific Gloves
G3 Sterile White Nitrile Hand Specific Gloves
Actual product may vary
G3 Sterile White Nitrile Hand Specific Gloves
Nitrile gloves feature a textured finish and provide enhanced sterile cleanroom protection for a wide range of contaminants in a Class 10/ISO 4 critical environment. Hand-specific pairs of Kimtech™ G3 Sterile White Nitrile gloves are appropriate for workers and technicians in medical device manufacturing, pharmaceuticals, or research labs.
Have Questions?
Catalog NumberSize
KLC568917.5
Catalog number KLC56891
Price (MYR)
1,478.00
CS
Add to cart
Size:
7.5
Price (MYR)
1,478.00
CS
Add to cart
Nitrile gloves feature a textured finish and provide enhanced sterile cleanroom protection for a wide range of contaminants in a Class 10/ISO 4 critical environment. Hand-specific pairs are appropriate for workers and technicians in medical device manufacturing, pharmaceuticals, or research labs.
  • White nitrile construction, validated to a Sterility Assurance Level (SAL) of 10-6, and cleanroom-ready packaging
  • Fully sterilized material features certified, high levels of contamination protection against particles, micro-organisms, viruses, cytotoxic drugs and chemical splash
  • Hand-specific, disposable and offer a high level of cleanliness
  • Textured fingertips enhance grip and tactile sensitivity for safer and more efficient processes
  • Beaded cuffs add strength and reduces the risk of tearing
  • Does not contain natural rubber latex, silicone or powder, reducing the risks of skin irritation
Specifications
Certifications/ComplianceASTM D5151, ASTM D412, ASTM D573, ASTM D3767, ASTM D3577, ASTM D3767, ASTM D6319, IEST-RP-CC005, ASTM D6978-05
Certifications/Compliance (Europe)EN ISO 374-1:2016; EN 374-4:2003; EN ISO 374-5:2016
Chemotherapeutic RatedYes
Cleanroom ClassClass 10 (ISO 4)
Accelerator FreeYes
AQL1.5
ColorWhite
DisposableSingle-use
Exterior FinishFully Textured
Finger Thickness (English)6 mil
Finger Thickness (Metric)0.15 mm
HandSpecific
Latex FreeYes
MaterialNitrile
Palm Thickness (English)5.1 mil
Palm Thickness (Metric)0.13 mm
PowderedPowder Free
Product TypeCleanroom Gloves
Size7.5
SterilitySterile
Thickness (English)6 mil
Thickness (Metric)0.15 mm
Model56891
SeriesKimtech™ G3 Sterile
Length (English)12 in.
Length (Metric)30.5 cm
Unit SizeCS