Kimtech™ G3 White Nitrile Gloves
Kimtech™ G3 White Nitrile Gloves
Actual product may vary
Kimtech™ G3 White Nitrile Gloves
Ambidextrous Kimtech™ G3 white nitrile gloves, size X-Large.
Have Questions?
Catalog number KLC56886
Price (MYR)
1,303.00
CS
Add to cart
Price (MYR)
1,303.00
CS
Add to cart
Pack of 1000 ambidextrous Kimtech™ G3 white nitrile gloves, size X-Large.
  • (Formerly HC61014).
  • Ideal for handling delicate equipment in microelectronics, semiconductor, optics and non-sterile pharmaceutical applications; when the highest levels of sensitivity, protection and performance are required.
  • The high quality nitrile gloves deliver seamless protection when and where it is needed and are suitable for ISO Class 3 or higher cleanroom environments.
  • The nitrile polymer material is designed for fit and reliability, with textured fingertips for improved grip and excellent water tightness with a low risk of pinholes.
  • The gloves are ambidextrous and incorporate a beaded cuff for added strength and ease of donning, so the wearer can simply grab and go without any fear of ripping the material.
  • Our non-sterile nitrile cleanroom gloves are also latex, silicone and powder-free.
  • The absence of natural rubber latex reduces the risk of TYPE 1 glove-associated reactions, protecting the wearer as well as the application.
Specifications
ColorWhite
DisposableSingle-use
Exterior FinishFingertip Textured
Finger Thickness (Metric)5.1 mm
HandAmbidextrous
Latex FreeYes
MaterialNitrile
PowderedPowder Free
SizeX-Large
SterilitySterile
Thickness (Metric)5.1 mm
Unit SizeCS