Pure G3 White Cleanroom Nitrile Gloves
Pure G3 White Cleanroom Nitrile Gloves
Actual product may vary
Pure G3 White Cleanroom Nitrile Gloves
White, non-sterile gloves offer a comfortable fit, smooth finish, dexterity and tactile sensitivity. Kimtech™ Pure G3 White Cleanroom Nitrile Gloves recommended for ISO Class 4 or higher cleanroom environments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
KLC56881Small
KLC56880X-Small
KLC56882Medium
KLC56883Large
Catalog number KLC56881
Price (MYR)
1,303.00
CS
Add to cart
Size:
Small
Price (MYR)
1,303.00
CS
Add to cart
  • Made of soft, 100% nitrile that resists oils and a broad range of chemicals
  • Double bagged with case liner, recommended for ISO Class 4 or higher cleanrooms
  • Chlorinated to remove powders used in manufacturing
  • Subjected to multiple deionized water rinses and packaged in a cleanroom environment
  • Proprietary processing used for lowest particle and ionic/elemental extractables
  • ESD characteristics
  • Tapered, beaded cuffs for reduced tearing and easy donning and doffing
  • Ambidextrous
  • White
Specifications
Certifications/ComplianceEN374-3:2003, IEST-RP-CC005 EN374-1:2003, AQL 1.5 / EN374-2, ISO16604, CE directive 89/686/EEC
Cleanroom ClassClass 1 (ISO 3)
Accelerator FreeYes
ColorWhite
DisposableSingle-use
Exterior FinishPartially Textured
Finger Thickness (English)6.2 mil
Finger Thickness (Metric)0.15 mm
HandAmbidextrous
Latex FreeYes
MaterialNitrile
Palm Thickness (English)5.1 mil
Palm Thickness (Metric)0.13 mm
PowderedPowder Free
Product TypeControlled Environment Gloves
SizeSmall
SterilityNon-sterile
Thickness (English)6.2 mil
Thickness (Metric)0.15 mm
Model56881
SeriesKimtech™ Pure G3
Length (English)12 in.
Length (Metric)30 cm
Unit SizeCS