Specifications
Product TypeGeneral Purpose Coveralls
SizeX-Large
GenderUnisex
MaterialMicroporous Film Laminate
ColorWhite
DisposableLimited-use
Ankle StyleElastic
Cuff StyleElastic
Neck StyleHooded
Hood TypeElastic, Respirator Fit
Closure TypeFront Zipper
Seam TypeSerged
SterilityNon-sterile
ResistanceLiquid, Particle, Anti-static, Chemicals, Bloodborne Pathogens
Certifications/ComplianceNFPA 99, ASTM F1670
ModelKleenGuard™ A40 Reflex™
No. of PocketsNone
For Use With (Application)Aviation, Pharmaceutical Manufacturing, General Manufacturing, Liquid Handling
Unit SizeCase