Polyethylene Oily Waste Cans
Polyethylene Oily Waste Cans
Actual product may vary
Polyethylene Oily Waste Cans
Designed for temporary storage of solvent, waste rags, and other materials subject to spontaneous combustion
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (English)ColorHeight (English)Width (English)
JUS933FL6 gal.Red16 in.16.5 in.
FIS19-038-4526 gal.Yellow16 in.16.5 in.
Catalog number JUS933FL
Price (MYR)
-
Capacity (English):
6 gal.
Color:
Red
Height (English):
16 in.
Width (English):
16.5 in.
Durable, fire-safe cans won't rust or dent and resist most chemical compounds.
  • Cans feature foot treadle that opens cover; self-closing lid
  • Choice of red or high-visibility yellow
Specifications
Capacity (Metric)23 L
Certifications/ComplianceFM, OSHA 29 CFR 1910.108, 20 CFR 1910.106
Closure TypeSelf Closing
Capacity (English)6 gal.
ColorRed
Dimensions (L x W x H)13.5 x 16.5 x 16 in. (34.29 x 41.6 x 41 cm)
Height (Metric)41 cm
Length (Metric)34.29 cm
Quantity1 Ea.
Width (Metric)41.6 cm
IncludesFoot lever
For Use With (Application)For temporary storage of solvent, waste rags, and other materials subject to spontaneous combustion
Width (English)16.5 in.
Length (English)13.5 in.
Height (English)16 in.
MaterialHDPE
Product TypeOily Waste Can
Unit SizeEA