General Purpose Swabs
General Purpose Swabs
Actual product may vary
General Purpose Swabs
Designed for industrial or general-purpose applications
Have Questions?
Catalog NumberLength (Metric)Length (Metric) TipQuantityTip Material
ITWTX80112.7 cm0.25 cm6 Bags/Cs.Polyester, Foam
Catalog number ITWTX801
Price (MYR)
-
Length (Metric):
12.7 cm
Length (Metric) Tip:
0.25 cm
Quantity:
6 Bags/Cs.
Tip Material:
Polyester, Foam
Polypropylene (100%) handle provides chemical compatibility
Complete thermal bond construction eliminates adhesive contamination
Packaged in a reclosable, silicone-free bag
Available in Polyester Honeycomb and Foam
Manufactured to exacting and consistent tolerances using high-precision automated processes
Lot coded for traceability and quality control
Orange-handle provides easy identification for use in less critical areas
Polyester 801 and 802
Honeycomb-type cell designed to maximize absorbency, capture large particles and dirt
Fabric “cells” engineered for pickup and application of adhesives, greases and liquids
Foam 803, 804 and 805
Polyurethane (100%) foam
Engineered for applying and absorbing adhesives, greases and liquids in less critical areas
Applications:
Removing and applying solders and fluxes
Cleaning confined spaces containing thick solutions (such as paints or inks)
Applying and removing lubricants, adhesives and solutions
Picking up powders and dust
Specifications
Quantity6 Bags/Cs.
Diameter (English) Handle0.205 in.
Length (English)5 in.
For Use With (Application)Removing and applying solders and fluxes, applying and removing lubricants, adhesives and solutions
Handle ColorOrange
Product TypeSwab
Diameter (Metric) Handle0.52 cm
Length (Metric)12.7 cm
Length (English) Handle4.03 in.
Length (Metric) Handle10.2 cm
MaterialPolyster
Length (English) Tip0.992 in.
Length (Metric) Tip0.25 cm
Width (Metric) Tip6.2 mm
Width (English) Tip0.5 in.
Handle MaterialPolypropylene
Tip StyleHoneycomb
Tip MaterialPolyester, Foam
Unit SizeCase