Microdenier Swabs
Microdenier Swabs
Actual product may vary
Microdenier Swabs
Made of microdenier material with complete thermal bond construction that eliminates adhesive contamination
Have Questions?
Catalog NumberLength (Metric)Tip StyleLength (Metric) HandleLength (Metric) Tip
ITWTX714MD12.7 cmFlat10.1 cm2.57 cm
Catalog number ITWTX714MD
Price (MYR)
-
Length (Metric):
12.7 cm
Tip Style:
Flat
Length (Metric) Handle:
10.1 cm
Length (Metric) Tip:
2.57 cm
Made of microdenier material with complete thermal bond construction that eliminates adhesive contamination
Nylon-free swab is compatible with QAC (quaternary ammonium chloride) cleaners and disinfectants
Material-100% polyester fabric
Enhanced absorbency and quick wicking performance
Offers increased lifting and capturing of particles as small as microbes
Chemical resistant and provides streak-free cleaning
Cleanroom laundered, providing ultra low levels of particles, NVRs (non volatile residues) and ions
Handle-100% virgin polypropylene
Autoclavable for use in sterile environments and processes
Packaged in a reclosable, silicone-free bag
Applications:
Applying and removing lubricants, adhesives and other solutions in critical clean environment
Scrubbing recessed areas
Removal of excess materials, debris
Cleaning intersecting surfaces and joints
Cleaning with compatible solutions and solvents
Picking up fine powders
Specifications
Diameter (English) Handle0.205 in.
Length (English)5 in.
For Use With (Application)Applying and removing lubricants, adhesives and other solutions in a critical clean environment, removal of excess materials, debris, cleaning intersecting surfaces and joints
Handle ColorLight Green
Product TypeSwab
Diameter (Metric) Handle0.52 cm
Length (Metric)12.7 cm
Length (English) Handle4 in.
Length (Metric) Handle10.1 cm
MaterialPolyester (100%)
Length (English) Tip1 in.
Handle TypePolypropylene, light green and easy grip handle
Length (Metric) Tip2.57 cm
Width (Metric) Tip12.7 mm
Width (English) Tip0.1 in.
AutoclavableAutoclavable
Handle MaterialPolypropylene
Tip StyleFlat
Tip MaterialMicrodenier Polyester
Unit SizeCase