TexQ™ Quaternary Ammonium Disinfectants
Actual product may vary
TexQ™ Quaternary Ammonium Disinfectants
One step cleaner and disinfectant
Have Questions?
Catalog NumberContainer Type
ITWTX650Bottle
Catalog number ITWTX650
Price (MYR)
-
Container Type:
Bottle
  • Dye/fragrance free
  • Effective against bacteria, virus and fungi
  • Available in a 22 oz. bottle or 1 gallon concentrate
  • EPA registered
Specifications
SterilitySterile
Certifications/ComplianceEPA
Container TypeBottle
For Use With (Application)Effective against a broad spectrum of Bacteria, Virus and Fungi and inhibits the growth of mold and mildew and their odors
Volume (English)22 oz.
Unit SizeCase