TechniCloth™ Nonwoven Dry Wipers, ISO Class 6/7
TechniCloth™ Nonwoven Dry Wipers, ISO Class 6/7
Actual product may vary
TechniCloth™ Nonwoven Dry Wipers, ISO Class 6/7
For ISO Class 6 and 7 applications
Have Questions?
Catalog NumberDimensions (L x W)
ITWTX6099 x 9 in. (22.86 x 22.86 cm)
Catalog number ITWTX609
Price (MYR)
2,206.00
CS
Add to cart
Dimensions (L x W):
9 x 9 in. (22.86 x 22.86 cm)
Price (MYR)
2,206.00
CS
Add to cart
For ISO Class 6 and 7 applications
Economical all-purpose nonwoven wipers made from a blend of 55% cellulose and 45% polyester—combine the absorbency of natural fiber with the cleanliness and strength of synthetic fiber.
  • Hydroentanglement process uses jets of water to blend fibers, eliminating need for chemical binders
  • Provide excellent bidirectional strength and very low solvent extractables levels
  • Offer superior wet and dry strength, solvent-compatibility and acid resistance
  • Will not scratch or abrade delicate precision components
Specifications
Product TypeDry Wipe
Length (English)9 in.
Length (Metric)22.9 cm
Width (English)9 in.
Width (Metric)22.9 cm
MaterialPolyester Cellulose
AutoclavableAutoclavable
Basis Weight69
Certifications/ComplianceISO 9001:2000, 13485:2003, 14001:2004 and OHSAS 18001:1999
Dimensions (L x W)9 x 9 in. (22.86 x 22.86 cm)
For Use With (Application)Designed for general purpose wiping where economy and cleanliness are of critical importance, ideal for applications where high absorbency and low extractables are prime considerations
Unit SizeCS