ThermaSeal™ 60 Wipers
Product Image
ThermaSeal™ 60 Wipers
100% continuous-filament, double-knit polyester fabric. These wipers are laser cut (thermally sealed) polyester, laundered in an ISO Class 4. Ideal for all purpose applications in critical cleanrooms and higher environments.
Have Questions?
Catalog NumberDimensions (L x W)
ITWTX2069B10 x 10 cm
Catalog number ITWTX2069B
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Dimensions (L x W):
10 x 10 cm
  • Offer good absorbency, wiping efficiency and abrasion resistance while maintaining chemical compatibility with standard solvents and alcohols
  • Wiping and cleaning surfaces, equipment and parts
  • Applying and removing lubricants, adhesives, residues and other solutions including disinfectants
  • Cleaning with solvents such as isopropyl alcohol (IPA), ethanol, acetone, and degreasers
  • Lining trays for holding, protecting, drying and storing of parts, equipment and devices
  • Appropriate for use with temperatures less than 400°F (205°C)
Specifications
MaterialPolyester
Dimensions (L x W)10 x 10 cm
Packaging Quantity2 Bags x 75 Wipes, 10 Bags/Cs.
Unit SizeCS