FlexiPump
FlexiPump
Actual product may vary
FlexiPump
The FlexiPump is a peristaltic dispensing pump for culture media preparation.
Have Questions?
Catalog number ISC561000
Price (MYR)
-
FlexiPump is a peristaltic pump with enhanced accuracy for the dispensing of culture media, agar and any type of diluent. Compact and ergonomic, it comes with a user-friendly interface.
  • Dispensing mode: Continuous, Dose, Multi-dose
  • Dose volume: from 50 μL to 99.9 L
  • Flow rate: Up to 1.25 L/min.
Specifications
Control TypeAnalog
Display TypeDigital
Enclosure RatingIP31
Flow Rate1.5 mL/min. to 1.25 L/min.
Motor Speed10 to 550 rpm
ReversibleNo
ModelFlexiPump
Voltage100/240 V
Unit SizeBX