Check Order Status
Scan 100
Actual product may vary
Scan 100
Manual colony counter
Have Questions?
Catalog number
ISC435000
Price (MYR)
-
Manual colony counter with LEDs Lighting
USB data transfer
Black/white background (100/240 V)
Specifications
Product Type
Colony Counter (Manual)
Display Type
Backlit LCD
Voltage
100/240 V
Unit Size
BX