BagLight PolySilk Blender Bags
BagLight PolySilk Blender Bags
Actual product may vary
BagLight PolySilk Blender Bags
BagLight PolySilk blender bags.
Have Questions?
Catalog NumberCapacity (Metric)
ISC132025400 mL
Catalog number ISC132025
Price (MYR)
-
Capacity (Metric):
400 mL
Sterile BagLight PolySilk blender bags without filter. Pack of 25.
  • Ideal for sample preparation.
  • Highly durable as they are able to withstand long blending times and avoid breakage from sample types known to cause problems.
  • Temperature resistance: from -40°C to 80°C.
  • 175 x 300 mm
  • Gamma ray treated.
Specifications
Capacity (Metric)400 mL
Volume Range (Metric)0 - 400 mL
MaterialPolymer
Unit SizeBX