BagFilter™ P Lateral Filter Bags
BagFilter™ P Lateral Filter Bags
Actual product may vary
BagFilter™ P Lateral Filter Bags
BagFilter™ P Lateral Filter Bags are non-woven side filters that are ideal for pipetting. Filtration is instantaneous without the risk of cross-contamination.
Have Questions?
Catalog NumberFor Use With (Equipment)
ISC111425Blender Bags
Catalog number ISC111425
Price (MYR)
-
For Use With (Equipment):
Blender Bags
BagFilter™ P Lateral Filter Bags are non-woven side filters that are ideal for pipetting. Filtration is instantaneous without the risk of cross-contamination.
  • The porosity of the filter: <250 μm
  • Compatible with any laboratory blender
  • Multilayer: Reinforced multicoated complex
  • Rigid and transparent
Specifications
Product TypeLateral Filter Bags
AutoclavableAutoclavable
Certifications/ComplianceISO 7218, ISO 6887, and FDA BAM (Bacteriological Analytical Manual)
Volume (Metric)400 mL
For Use With (Application)Microbiological Analyses
For Use With (Equipment)Blender Bags
Pore Size<250 μm
No. per Pack500 Pk.
Unit SizePK