Check Order Status
Diaphragm Vacuum Pump Service and Maintenance Kit
Actual product may vary
Diaphragm Vacuum Pump Service and Maintenance Kit
Service and Maintenance Kits comprised of replacement seals and wearable parts for various models of diaphragm pumps.
Have Questions?
Catalog Number
For Use With
ILM402008
Pump Models MP / MPC 101 Z, 201 E
Catalog number
ILM402008
Price (MYR)
-
For Use With:
Pump Models MP / MPC 101 Z, 201 E
Available for a range of diaphragm vacuum pump models
Provided individually
Specifications
For Use With
Pump Models MP / MPC 101 Z, 201 E
Includes
Diaphragm, Valves And O-Rings.
Quantity
1 Pack
Type
Service Kit