Magnetic Hotplate Stirrer, Ambient to 120°C, Aluminum Alloy
Product Image
Magnetic Hotplate Stirrer, Ambient to 120°C, Aluminum Alloy
Stirring Range: 0 to 1000 rpm, Shape: Rectangle, Size: 10.6 x 19.5 in., Plug Type: UK-Style
Have Questions?
Catalog NumberPlug Type
IKA3692602UK
Catalog number IKA3692602
Price (MYR)
45,048.00
EA
Add to cart
Plug Type:
UK
Price (MYR)
45,048.00
EA
Add to cart
Digital magnetic hotplate stirrer, designed for synchronous heating and stirring. The magnetic coil technology provides noiseless and consistent stirring on all positions. The surface temperature is infinitely adjustable up to 120 °C, producing a maximum medium temperature of 70 °C (depending on the used vessel). Speed remains constant, even when load changes.
  • Homogeneous temperature distribution across the heating plate
  • Adjustable reverse operation
  • Eco-mode for a low self-warming of the surface
  • Wear free magnetic coils
  • Error code display
  • Easy operation with touch keypad
  • Suitable for continuous operation
  • Speed range: 0 - 1000 rpm
Specifications
Top Plate MaterialAluminum Alloy
Voltage100/115/230 V
Stirring FeaturesSynchronous heating and stirring
ModelRT 15
Size10.6 x 19.5 in.
Height (Metric)60 mm
Temperature (Metric)Ambient to 120°C
Ambient Temperature5°C to 40°C
Frequency50/60 Hz
Width (Metric)280 mm
Wattage620 W
Weight (English)23.1 lb.
Weight (Metric)10.5 kg
TypeStirring Hotplate
ShapeRectangle
Plug TypeUK
No. of Positions15
Max. Container Size0.4 L
Stirring Range0 to 1000 rpm
Warranty24 Months
Certifications/ComplianceDIN EN 60529
DisplayLED
Unit SizeEA