Flat bouffant caps provide protection against dirt, grime, and particulates in non-hazardous environments. Available in blue or white in a variety of sizes.
Have Questions?
Change view
Catalog Number
Size
Color
IENS110NW-24B
24 in.
Blue
IENS110NW-24
24 in.
White
2 Options
Catalog number IENS110NW-24B
Price (MYR)
363.00
CS
Add to cart
Size:
24 in.
Color:
Blue
Price (MYR)
363.00
CS
Add to cart
Provide a particulate barrier and helps prevent hair from contaminating a product or work environment
Low-linting, lightweight cover restrains hair and offers great ventilation
Designed to protect against dirt, grime, and particulates in non-hazardous environments
Flat style
Latex-free elastic
Possible applications include food processing, construction, pharmaceuticals, environmental cleanup, and more
Specifications
Product TypeBouffant
Size24 in.
MaterialPolypropylene
ColorBlue
DisposableSingle-use
ModelS110NW
Diameter (English)24 in.
For Use With (Application)Food Processing, Construction, Pharmaceuticals, Environmental Cleanup