Check Order Status
VHP Union Assembly
Actual product may vary
VHP Union Assembly
Zero dead volume unions ensure accurate analysis
Have Questions?
Catalog Number
Diameter (Metric) Outer
Swept Volume
HYPU-435
0.25 mm
0.025 μL
Catalog number
HYPU-435
Price (MYR)
623.00
EA
Add to cart
Diameter (Metric) Outer:
0.25 mm
Swept Volume:
0.025 μL
Price (MYR)
623.00
EA
Add to cart
Precision machined from 316 stainless steel to withstand high pressures
Securely join fluid pathway components Holds tight connection even through vibration
VHP ZDV union assembly
10-32 coned for 1/16 in. OD tubing
Max pressure 20,000 psi (1380 bar)
Specifications
Type
Union
Diameter (English) Inner
0.01 in.
Diameter (Metric) Outer
0.25 mm
Pressure Rating
10000 psi
Swept Volume
0.025 μL
Material
Stainless Steel
For Use With (Equipment)
1/16 in. OD Tubing