Check Order Status
Male-Threaded Nuts
Actual product may vary
Male-Threaded Nuts
Combine with Super Flangeless™ ferrule for a high pressure holding flat-bottom fitting assembly
Have Questions?
Catalog Number
For Use With (Equipment)
Length (English)
Material
Width (English)
HYPU-400X
1/16 in. O.D. Tubing
0.4 in.
Stainless Steel
0.25 in.
Catalog number
HYPU-400X
Price (MYR)
838.00
PK
Add to cart
For Use With (Equipment):
1/16 in. O.D. Tubing
Length (English):
0.4 in.
Material:
Stainless Steel
Width (English):
0.25 in.
Price (MYR)
838.00
PK
Add to cart
Combine with Super Flangeless™ ferrule for a high pressure holding flat-bottom fitting assembly
Eliminates tubing twist that can loosen connections
Resistant to loosening through vibration
Specifications
Diameter (English) Outside
0.0625 in.
Length (English)
0.4 in.
Width (English)
0.25 in.
For Use With (Equipment)
1/16 in. O.D. Tubing
Material
Stainless Steel
Type
Nut