Check Order Status
High-Pressure, Zero Dead Volume Unions
Actual product may vary
High-Pressure, Zero Dead Volume Unions
PEEK ZDV unions provide accurate results
Have Questions?
Catalog Number
Diameter (English) Inner
Swept Volume
HYPP-742
0.01 in.
0.071 μL
Catalog number
HYPP-742
Price (MYR)
666.00
EA
Add to cart
Diameter (English) Inner:
0.01 in.
Swept Volume:
0.071 μL
Price (MYR)
666.00
EA
Add to cart
Inert PEEK fluid path
10-32 coned configuration
Specifications
Type
Union
Diameter (English) Inner
0.01 in.
Pressure Rating
6000 psi
Swept Volume
0.071 μL
Quantity
Each
Material
PEEK
For Use With (Equipment)
1/16 in. OD Tubing