High-Pressure Crosses
High-Pressure Crosses
Actual product may vary
High-Pressure Crosses
PEEK High-Pressure Unions, Tees, and Crosses hold up to 5000 psi and include FINGERTIGHT III fittings. Fittings work with any 1/16 in. OD tubing.
Have Questions?
Catalog NumberDiameter (English) Inner
HYPP-7290.02 in.
Catalog number HYPP-729
Price (MYR)
1,086.00
EA
Add to cart
Diameter (English) Inner:
0.02 in.
Price (MYR)
1,086.00
EA
Add to cart
Specifications
TypeCross
Diameter (English) Inner0.02 in.
Pressure Rating3500 psi
Swept Volume0.721 μL
MaterialPEEK
ColorBlack
For Use With (Equipment)1/16 in. O.D. Tubing
Unit SizeEA