Tee
Tee
Actual product may vary
Tee
Combine multiple fluid streams into one under high operating pressures
Have Questions?
Catalog NumberMaterial
HYPP-727PEEK
Catalog number HYPP-727
Price (MYR)
724.00
EA
Add to cart
Material:
PEEK
Price (MYR)
724.00
EA
Add to cart
  • Securely join fluid pathway components Holds tight connection even through vibration
  • High-pressure tee
  • Max pressure 3500 psi (241 bar)
  • 0.020 in. thru-hole diameter
  • Swept volume of 0.57 μL
Specifications
TypeTee
Diameter (English) Inner0.02 in.
QuantityEach
MaterialPEEK
For Use With (Equipment)1/16 in. OD Tubing
Unit SizeEA