Fingertight Two-Piece, Double-Wing, Natural PEEK, 1/16 in. OD Tubing, 10-32 Coned
Fingertight Two-Piece, Double-Wing, Natural PEEK, 1/16 in. OD Tubing, 10-32 Coned
Actual product may vary
Fingertight Two-Piece, Double-Wing, Natural PEEK, 1/16 in. OD Tubing, 10-32 Coned
Withstands up to 6000 psi.
Have Questions?
Catalog NumberMaterial
HYPF-300XPEEK
Catalog number HYPF-300X
Price (MYR)
1,179.00
PK
Add to cart
Material:
PEEK
Price (MYR)
1,179.00
PK
Add to cart
Get the convenience of nut and ferrule built into one fitting. By using one-piece fittings there is no need to worry about the ferrule getting stuck in a receiving port.
Specifications
TypeTwo Piece Fingertight Double Winged Head Nut; 10 to 32 Coned
IncludesFerrule
MaterialPEEK
For Use With (Equipment)For 0.06 in. O.D. Tubing
Unit SizePK