One-Piece, High-Pressure Fingertight Fittings: PEEK
One-Piece, High-Pressure Fingertight Fittings: PEEK
Actual product may vary
One-Piece, High-Pressure Fingertight Fittings: PEEK
Fingertight coned fittings can withstand up to 9000 psi.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (English)Pressure Rating
HYPF-130X1.03 in.6000 psi
HYPF-120X0.82 in.5000 psi
Catalog number HYPF-130X
Price (MYR)
1,179.00
PK
Add to cart
Length (English):
1.03 in.
Pressure Rating:
6000 psi
Price (MYR)
1,179.00
PK
Add to cart
Fingertight coned fittings can withstand up to 9000 psi.
Fingertight fittings offer the convenience of making a tubing connection without the need for wrenches or specialized tools when tightening the fitting into place
  • Pressures up to 7000psi (483 bar)
  • Temperatures up to 125°C
  • Fingertights for 1/16 in. OD and smaller tubing
  • Replaceable Ferrules for two-piece versions
Specifications
TypeFingertight Fitting
Length (English)1.03 in.
Pressure Rating6000 psi
MaterialPEEK
ColorNatural
For Use With (Equipment)0.0625 in. O.D. Tubing
Unit SizePK