Gas Regulators for Reacti-Vap™ Evaporators
Product Image
Gas Regulators for Reacti-Vap™ Evaporators
Use these gas regulators to deliver the right amount of gas for sample evaporation using the Thermo Scientific™ ReactiVap™ Evaporator.
Have Questions?
Catalog NumberDescription
HYP60181-626Gas Regulator, 1/4in. Tube Fitting
Catalog number HYP60181-626
Price (MYR)
4,648.00
EA
Add to cart
Description:
Gas Regulator, 1/4in. Tube Fitting
Price (MYR)
4,648.00
EA
Add to cart
Deliver the right amount of gas to your Thermo Scientific™ ReactiVap™ Evaporator using these gas regulators. The Reacti-Vap Evaporator delivers pressurized gas to easily and efficiently evaporate samples for gas chromatography (GC) analysis. Choose between gas regulators with a 1/8 inch or a 1/4 inch tube fitting.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionGas Regulator, 1/4in. Tube Fitting
For Use WithReacti-Vap Evaporators
Product TypeGas Regulator (1/4 in. Tube Fitting)
Unit SizeEA