Check Order Status
TriPlus RSH Autosampler Fixed Needle Syringe, 10μL
TriPlus RSH Autosampler Fixed Needle Syringe, 10μL
Thermo Scientific™ TriPlus RSH Autosampler Fixed Needle Syringes are accurate and reproducible over a range of volumes.
Have Questions?
Change view
Catalog Number
Needle Gauge
Length (Metric) Needle
HYP365D0321
26s G
85 mm
365D0321
26s G
85 mm
2 Options
Catalog number
HYP365D0321
Price (MYR)
1,112.00
EA
Add to cart
Needle Gauge:
26s G
Length (Metric) Needle:
85 mm
Price (MYR)
1,112.00
EA
Add to cart
Thermo Scientific™ TriPlus RSH Autosampler Fixed Needle Syringes are accurate and reproducible over a range of volumes.
Borosilicate glass with metal flanges
Needles and plungers are stainless steel
Field-proven syringe design
Specifications
Description
Splitless and OC
Needle Gauge
26s G
Product Type
Fixed Needle Syringe
Volume (Metric)
10 μL
Length (Metric) Needle
85 mm
Needle Point Style
Cone