SilTite™ Ferrules
SilTite™ Ferrules
Actual product may vary
SilTite™ Ferrules
Thermo Scientific™ SilTite Ferrules are for use with capillary column connectors.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)
HYP290MF2290.25 mm
290MF2290.25 mm
290MF2300.32 mm
HYP290MF2300.32 mm
Catalog number HYP290MF229
Price (MYR)
873.00
PK
Add to cart
Diameter (Metric):
0.25 mm
Price (MYR)
873.00
PK
Add to cart
Thermo Scientific™ SilTite Ferrules are for use with capillary column connectors.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionFor 0.25 mm I.D. Capillary; Pk. of 10
Diameter (Metric)0.25 mm
For Use With (Equipment)Fused Silica Capillary GC Columns
MaterialSilTite Metal
TypeFerrule
Unit SizePK