TRACE™ TR-1701 GC Columns
TRACE™ TR-1701 GC Columns
Actual product may vary
TRACE™ TR-1701 GC Columns
Use mid-polarity columns with alternative selectivity with Thermo Scientific™ TRACE™ TR-1701 GC Columns.
Have Questions?
Catalog NumberLength (Metric)Diameter (Metric) InnerFilm Thickness
HYP260Q142P30 m0.25 mm0.25 μm
Catalog number HYP260Q142P
Price (MYR)
4,786.00
EA
Add to cart
Length (Metric):
30 m
Diameter (Metric) Inner:
0.25 mm
Film Thickness:
0.25 μm
Price (MYR)
4,786.00
EA
Add to cart
Use mid-polarity columns with alternative selectivity with Thermo Scientific™ TRACE TR-1701 GC Columns.
  • Mid-polarity phase, 14% Cyanopropylphenyl polysiloxane
  • Low bleed even at a high operating temperature
  • Excellent starting point for method development
  • Suitable for a wide variety of applications
  • Similar to DB-1701, Rtx-1701, HP-1701, BP10, OV-1701, 007-1701, CP-Sil 19 CB
Recommended for:
  • Pesticides
  • PCB's
  • PAH's
  • Organic Acids
  • Drugs
  • Steroids
  • EPA 608, 8081
Specifications
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
Product LineTRACE
Stationary PhaseTrace™ TR-1701
USP TypeG46
Max. Temperature280/300°C
For Use With (Application)GC/MS for Pesticides, PCBs, PAHs, and Organic Acids, GC/MS for Drugs and Steroids, GC/MS for EPA 608, 8081
Film Thickness0.25 μm
Length (Metric)30 m
PolarityMid-Polar
Description30 m x 0.25 mm x 0.25 μm
Unit SizeEA