HyPURITY™ Cyano HPLC Columns
HyPURITY™ Cyano HPLC Columns
Actual product may vary
HyPURITY™ Cyano HPLC Columns
HyPURITY HPLC Columns are highly characterized, tested, and validated.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)
HYP22805-254630250 mm
22805-254630250 mm
Catalog number HYP22805-254630
Price (MYR)
6,516.00
EA
Add to cart
Length (Metric):
250 mm
Price (MYR)
6,516.00
EA
Add to cart
Thermo Scientific™ HyPURITY™ Cyano HPLC Columns are based on the HyPURITY 190Å silica—the highest quality silica available and essentially metal-free with exceptionally stablity at high and low pH. The rugged and robust manufacturing process ensures reproducibility and optimum quality, batch after batch. Within the manufacturing process, extra care is taken to ensure an extensively homogenous silica surface. This surface is a key precursor in the preparation of uniform bonded phases. Cyano functionality is generally less hydrophobic and therefore compounds are less retained compared to other alkyl phases, though it does have strong dipole interaction. Additionally, cyano can be operated under either reversed-phase or normal phase / HILIC conditions.
Specifications
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
For Use WithSVL™ Stirrer Bearing
Length (Metric)250 mm
Particle Size5 μm
Pore Size190 Å
Surface Area200 m²/g
USP TypeL10
Stationary PhaseCyano (CN)
Packing MaterialSilica
Carbon Load4%
Column TypeMixed Mode
EndcappedYes
PhaseReversed Phase, Normal Phase, Polar Selectivity
MaterialGlass
TypeTorion Body
Unit SizeEA