MAbPac™ Reversed Phase HPLC Columns
MAbPac™ Reversed Phase HPLC Columns
Actual product may vary
MAbPac™ Reversed Phase HPLC Columns
Use the new Thermo Scientific™ MAbPac™ RP columns for high-performance reversed-phase (RP) HPLC and LC/MS characterization of monoclonal antibodies (mAbs), fragments, variants, antibody drug conjugates (ADCs), PEGylated proteins, and bispecific proteins.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Stationary Phase
HYP088644100 mm3 mmPhenyl (Ph)
88644
also known as 088644
100 mm3 mmPhenyl (Ph)
Catalog number HYP088644
Price (MYR)
7,510.00
EA
Add to cart
Length (Metric):
100 mm
Diameter (Metric):
3 mm
Stationary Phase:
Phenyl (Ph)
Price (MYR)
7,510.00
EA
Add to cart
Use the new Thermo Scientific™ MAbPac™ Reversed Phase columns for high-performance reversed-phase (RP) HPLC and LC/MS characterization of monoclonal antibodies (mAbs), fragments, variants, antibody drug conjugates (ADCs), PEGylated proteins, and bispecific proteins. These columns have excellent resolution, long column lifetime, and low carry-over. The surface chemistry, particle size, and pore size, have been optimized for high resolution separations.
Compatible with organic solvent used in HPLC and MS under RP conditions
  • Resin is based on a hydrophobic supermacroporous 4μm polymer particles
  • Stable at extreme pH (0 to 14) and high temperature (up to 110 °C)
Efficient with low carry over
  • 1,500Å large pore size polymeric resin
  • Efficient separation of protein molecules with very low carry over
Designed for reversed-phase biomolecule LC and LC/MS separations
  • Superior resolution power for monoclonal antibodies and related substances
  • High efficiency with low carry-over
  • Excellent MS compatibility
  • Wide operating pH range: 0 to 14
  • High temperature stability: up to 110°C
  • High throughput
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column FormatAnalytical Column
Diameter (Metric)3 mm
Length (Metric)100 mm
Max. Pressure4000 psi (275 bar)
Particle Size4 μm
pH0 to 14
Pore Size1500 Å
QuantityEach
Temperature<110°C
Stationary PhasePhenyl (Ph)
Product LineMAbPac
Packing MaterialDVB (Divinylbenzene) Polymer
Column TypeReversed Phase
PhaseReversed Phase
TechniqueBio-LC
Green FeaturesResponsibly packaged
Unit SizeEA