Hot Plate, 38°C to 371°C, Aluminum
Hot Plate, 38°C to 371°C, Aluminum
Actual product may vary
Hot Plate, 38°C to 371°C, Aluminum
Shape: Square; Max. Sample Load: 18.2 kg.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberShapeSizeVoltageWattage
HPA2240MQRectangle12 x 24 in.240 V3200 w
HPA2230MQSquare12 x 12 in.240 V1600 w
Catalog number HPA2240MQ
Price (MYR)
11,204.00
EA
Add to cart
Shape:
Rectangle
Size:
12 x 24 in.
Voltage:
240 V
Wattage:
3200 w
Price (MYR)
11,204.00
EA
Add to cart
For constant temperature applications where requiring even heat (to 371°C)
  • Large, aluminum heating surface and thermostatic controls for superior temperature uniformity and stability
  • Thermostatic temperature control provides excellent stability
  • Safe and reliable construction
  • Cycle light indicates when power is being supplied to heating element
  • Stainless-steel case provides optimal strength for heavy loads
  • Epoxy-painted surface increases chemical resistance in corrosive environments
  • Large 12 x 12 in. (30.5 x 30.5 cm) and 12 x 24 in. (30.5 x 60.9 cm) top plate surfaces accommodate up to 40 lb. (18.2 kg) loads
Specifications
Top Plate MaterialAluminum
Temperature (English)100°F to 700°F
Size12 x 24 in.
Temperature (Metric)38°C to 371°C
Depth (English)13 in.
Depth (Metric)33.02 cm
Height (English)6.13 in.
Weight (English)40 lb.
Height (Metric)15.57 cm
Weight (Metric)18.14 kg
Wattage3200 w
Amperage13.3 A
Electrical Requirements240 V, 50/60 Hz
Frequency50/60 Hz
Max. Sample Load18 kg
ShapeRectangle
Voltage240 V
TypeHot Plate
Certifications/ComplianceUL Listed
Warranty3 Years
Dimensions (L x W x H)12 in. x 24 in.
Unit SizeEA