Aluminum-Top Hotplates
Aluminum-Top Hotplates
Actual product may vary
Aluminum-Top Hotplates
Cast-aluminum top for maximum heat transfer, strength and corrosion resistance.
Have Questions?
Catalog NumberVoltage
HPA1910MQ240 V
Catalog number HPA1910MQ
Price (MYR)
2,331.00
EA
Add to cart
Voltage:
240 V
Price (MYR)
2,331.00
EA
Add to cart
Cast-aluminum top for maximum heat transfer, strength and corrosion resistance.
  • Equipped with thermostatic temperature control
  • Baffle-vented welded stainless-steel case
  • Recommended for use with glass vessels only
Warranty: Three years
Certifications: UL listed (120V model)
Specifications
Top Plate MaterialAluminum
Size6.25 x 6.25 in.
Temperature (Metric)38°C to 371°C
Height (English)4.38 in.
Length (English)7.63 in.
Width (English)6.63 in.
Height (Metric)11.1 cm
Length (Metric)19.4 cm
Width (Metric)16.8 cm
ShapeSquare
Voltage240 V
TypeHot Plate
Certifications/ComplianceUL Listed, Certificate of Analysis (COA), Certificate of Conformance (COC)
Dimensions (L x W x H)6.25 in. x 6.25 in.
Unit SizeEA